missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CCL11 (Eotaxin), Animal-Free Recombinant Protein, PeproTech®
Click to view available options
Quantity:
250 μg
5 μg
Unit Size:
250µg
5µg
Description
Recombinant Human Eotaxin is an 8.3 kDa protein containing 74 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
This gene is one of several Cys-Cys (CC) cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for eosinophils, but not mononuclear cells or neutrophils. This eosinophil specific chemokine assumed to be involved in eosinophilic inflammatory diseases such as atopic dermatitis, allergic rhinitis, asthma and parasitic infections.
Specifications
Specifications
| Accession Number | P51671 |
| For Use With (Application) | Functional Assay |
| Format | Lyophilized |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 6356 |
| Molecular Weight (g/mol) | 8.3 kDa |
| Name | Human CCL11 (Eotaxin), Animal-Free |
| Purification Method | HPLC, SDS-PAGE |
| Quantity | 250 μg |
| Regulatory Status | RUO |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction