missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL14/HCC-1/HCC-3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3241-SP
This item is not returnable.
View return policy
Description
CCL14/HCC-1/HCC-3 Monoclonal specifically detects CCL14/HCC-1/HCC-3 in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| CCL14/HCC-1/HCC-3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 14, CCL14, chemokine (C-C motif) ligand 14, Chemokine CC-1/CC-3, chemokine CC-3, CKB1, FLJ16015, HCC1, HCC-1, HCC-1(1-74), HCC-1/HCC-3, HCC3, HCC-3, hemofiltrate CC chemokine 1, MCIF, member 14, NCC2CC-1, NCC-2CKb1, new CC chemokine 2, SCYA14CC-3, Small-inducible cytokine A14 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6358 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Neutralization, ELISA | |
| 256409 | |
| Western Blot 1 ug/mL, Neutralization 0.5-2.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| NP_116738 | |
| CCL14 | |
| E. coli-derived recombinant human CCL14/HCC-1/HCC-3 Thr20-Asn109 Accession # NP_116738 | |
| 25 μg | |
| Primary | |
| Detects human CCL14/HCC-1/HCC-3 in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human CCL1, 2, 3, 5, 7, 8, 11, 13, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, recombinant mouse CCL2, 3, 4, 6, 7, 9, 11, 12, 19, 20, 21, 22, 24, 25, or recombinant rat CCL20 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction