missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL14a/HCC-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF-324-SP
This item is not returnable.
View return policy
Description
CCL14a/HCC-1 Polyclonal specifically detects CCL14a/HCC-1 in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| CCL14a/HCC-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CCL14 | |
| E. coli-derived recombinant human CCL14a/HCC-1 Thr20-Asn93 Accession # Q16627 | |
| 25 μg | |
| Primary | |
| Detects human CCL14a/HCC-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots (non-reducing conditions), less than 5% cross-reactivity with recombinant human (rh) MCP-2, rhMCP-3, recombinant mouse (rm) C10, and rmMIP-1 beta is observed. | |
| Human | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.15-0.75 ug/mL | |
| Q16627 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6358 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction