missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL14a/HCC-1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF-324-PB
This item is not returnable.
View return policy
Description
CCL14a/HCC-1 Polyclonal specifically detects CCL14a/HCC-1 in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| CCL14a/HCC-1 | |
| Polyclonal | |
| Unconjugated | |
| Q16627 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6358 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.15-0.75 ug/mL | |
| CCL14 | |
| E. coli-derived recombinant human CCL14a/HCC-1 Thr20-Asn93 Accession # Q16627 | |
| 100 μg | |
| Primary | |
| Detects human CCL14a/HCC-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots (non-reducing conditions), less than 5% cross-reactivity with recombinant human (rh) MCP-2, rhMCP-3, recombinant mouse (rm) C10, and rmMIP-1 beta is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction