missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL17/TARC Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF364-SP
This item is not returnable.
View return policy
Description
CCL17/TARC Polyclonal specifically detects CCL17/TARC in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Specifications
| CCL17/TARC | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARC, TARCA-152E5.3, Thymus and activation-regulated chemokine | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6361 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.3-1.5 ug/mL | |
| Q92583 | |
| CCL17 | |
| E. coli-derived recombinant human CCL17/TARC (R&D Systems, Catalog # 364-DN) Ala24-Ser94 Accession # Q92583 | |
| 25 μg | |
| Primary | |
| Detects human CCL17/TARC in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse TARC is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction