missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL2/JE/MCP-1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC279G-025
This item is not returnable.
View return policy
Description
CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| CCL2/JE/MCP-1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6347 | |
| Human | |
| Ascites |
| Flow Cytometry | |
| 24822 | |
| Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
| P13500 | |
| CCL2 | |
| E. coli-derived recombinant human CCL2/JE/MCP-1 Accession # P13500 | |
| 25 Tests | |
| Primary | |
| Detects human CCL2/JE/MCP-1 in Western blots. Does not cross-react with recombinant human CCL1, 3, 4, 5, 7, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, recombinant mouse CCL1, 2, 3, 4, 5, 6, 7, 9,10, MIP-1 gamma , 11, 12, 17, 19, 20, 22, 24, 25, or recombinant rat CCL20. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction