missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCP-1/CCL2 Human Uncoated ELISA Kit with Plates
Specifically engineered for accurate and precise measurement of human CCL2 protein levels from samples including serum, plasma, and cell culture supernatants.
€ 242.00 - € 782.00
Specifications
Accession Number | P13500 |
---|---|
Assay Range | 7-1,000 pg/mL |
Assay Sensitivity | 7 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Alert:
Contains Azide
Product Code | Brand | Quantity | Gene Symbol | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Gene Symbol | Price | Quantity & Availability | ||||
15511147
|
Invitrogen™
88-7399-76 |
20 x 96 Tests | GCP-1,GCP1,IL8,LECT,LUCT,LYNAP,MDNCF,MONAP,NAF,NAP-1,NAP1 |
€ 782.00
Pack of 20 |
In Stock
Log in to see stock. |
||||
15521147
|
Invitrogen™
88-7399-77 |
20 x 96 Tests | IFN-g,Ifg |
€ 738.00
Pack of 20 |
In Stock
Log in to see stock. |
||||
15531147
|
Invitrogen™
88-7399-86 |
10 x 96 Tests | GCP-1,GCP1,IL8,LECT,LUCT,LYNAP,MDNCF,MONAP,NAF,NAP-1,NAP1 |
€ 544.00
Pack of 10 |
In Stock
Log in to see stock. |
||||
15541147
|
Invitrogen™
88-7399-88 |
10 x 96 Tests | IFN-g,Ifg |
€ 512.00
Pack of 10 |
In Stock
Log in to see stock. |
||||
15501147
|
Invitrogen™
88-7399-22 |
2 x 96 Tests | GCP-1,GCP1,IL8,LECT,LUCT,LYNAP,MDNCF,MONAP,NAF,NAP-1,NAP1 |
€ 242.00
Pack of 2 |
Estimated Shipment: 02-05-2024
Log in to see stock. |
||||
Description
Human CCL2 (MCP-1) ELISA kit (with or without high-affinity binding microwell plates) contains the necessary reagents, buffers and diluents for performing quantitative enzyme linked immunosorbent assays (ELISA)Specifications
P13500 | |
7 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Enzyme: Pre-titrated Streptavidin-HRP Concentrate (100X), Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86) | |
RUO | |
2°C to 8°C | |
Human | |
24 hr. 30 min. |
7-1,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
6347 | |
HRP | |
Plasma, 100 μL; Serum, 100 μL; Supernatant, 100 μL | |
Monocyte chemotactic protein 1 | |
1 hr. 45 min. |
For Research Use Only