missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL22/MDC Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC3361R-100UG
This item is not returnable.
View return policy
Description
CCL22/MDC Monoclonal specifically detects CCL22/MDC in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| CCL22/MDC | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O00626.1 | |
| CCL22 | |
| E. coli-derived recombinant human CCL22/MDC Gly25-Gln93 Accession # O00626.1 | |
| RUO | |
| 6367 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 57203 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDC, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CCL22/MDC in direct ELISAs and Western blots. In direct ELISAs, does not cross-react with recombinant cotton rat CCL3, 4, 5, recombinant human CCL1, 2, 3, 4, 5, 7, 8, 9/10/MIP-1 gamma , 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 23, 24, 25, 26, 27, 28, recombinant mouse CCL1, 2, 3, 4, 5, 6, 7, 8, 9/10/MIP-1 gamma , 11, 12, 17, 19, 21, 22, 24, 25, 27, 28, or recombinant rat CCL20. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction