missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CCL3 (P10147, 24 a.a. - 92 a.a.) Partial Recombinant Protein

Product Code. 16211960
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16211960 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16211960 Supplier Abnova™ Supplier No. P3651.20ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Func, SDS-PAGE

Macrophage inflammatory protein-1 is a so-called monokine that is involved in the acute inflammatory state in the recruitment and activation of polymorphonuclear leukocytes (Wolpe et al., 1988 [PubMed 3279154]). Sherry et al. (1988) [PubMed 3058856] demonstrated 2 protein components of MIP1, called by them alpha and beta.[supplied by OMIM]

Sequence: SLAADTPTACCFSYTSRQIPQNFIADYFETSSQCSKPGVIFLTKRSRQVCADPSEEWVQKYVSDLELSA

Specifications

Accession Number P10147
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 6348
Molecular Weight (g/mol) 8kDa
Name CCL3 (Human) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Ion exchange column and HPLC reverse phase column
Quantity 20 μg
Immunogen SLAADTPTACCFSYTSRQIPQNFIADYFETSSQCSKPGVIFLTKRSRQVCADPSEEWVQKYVSDLELSA
Storage Requirements Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg (1 EU/μg)
Gene Alias G0S19-1/LD78ALPHA/MIP-1-alpha/MIP1A/SCYA3
Common Name CCL3
Gene Symbol CCL3
Biological Activity Determined by its ability to chemoattract human monocytes using a concentration range of 1.0-10.0ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.