missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL5/RANTES Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC278N-100UG
This item is not returnable.
View return policy
Description
CCL5/RANTES Monoclonal specifically detects CCL5/RANTES in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| CCL5/RANTES | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| chemokine (C-C motif) ligand 5, D17S136Enormally T-expressed, and presumably secreted, EoCP, Eosinophil chemotactic cytokine, RANTES, SISd, SIS-delta, small inducible cytokine A5 (RANTES), small inducible cytokine subfamily A (Cys-Cys), member 5, Small-inducible cytokine A5, T cell-specific protein P228, T-cell specific protein p288, TCP228T-cell-specific protein RANTES | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CCL5/RANTES in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with other chemokines, including recombinant human CCL2, 3, 4, recombinant mouse (rm) CCL3, and rmCCL4. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 21445 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CCL5 | |
| E. coli-derived recombinant human CCL5/RANTES | |
| RUO | |
| 6352 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction