missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL7/MCP-3/MARC Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC282R-100UG
This item is not returnable.
View return policy
Description
CCL7/MCP-3/MARC Monoclonal specifically detects CCL7/MCP-3/MARC in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| CCL7/MCP-3/MARC | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q7Z7Q8 | |
| CCL7 | |
| E. coli-derived recombinant human CCL7/MCP-3/MARC Gln34-109 Accession # Q7Z7Q8 | |
| RUO | |
| 6354 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 36320 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| C-C motif chemokine 7, chemokine (C-C motif) ligand 7, FIC, MARC, MCP-3, MCP3Monocyte chemotactic protein 3, MCP-3Small-inducible cytokine A7, MGC138465, Monocyte chemoattractant protein 3, NC28SCYA6, SCYA7MGC138463, small inducible cytokine A7 (monocyte chemotactic protein 3) | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CCL7/MCP-3/MARC in ELISAs and Western blots. In ELISAs, no cross-reactivity with recombinant human (rh) CCL2/MCP-1, rhCCL8/MCP-2, or rhCCL13/MCP-4 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction