missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR1 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1451R-100UG
This item is not returnable.
View return policy
Description
CCR1 Monoclonal specifically detects CCR1 in Human samples. It is validated for Flow Cytometry.
Specifications
| CCR1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
| Mouse myeloma cell line NS0-derived recombinant human CCR1 Met1-Phe355 Accession # P32246 | |
| 100 μg | |
| 1230 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 984303 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P32246 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CCR1 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction