missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR1 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB145R-100UG
This item is not returnable.
View return policy
Description
CCR1 Monoclonal specifically detects CCR1 in Human samples. It is validated for Flow Cytometry.
Specifications
| CCR1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P32246 | |
| CCR1 | |
| Swiss 3T3 mouse cell line transfected with human CCR1 Accession # P32246 | |
| RUO | |
| 1230 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 53504 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CCR1. Reacts specifically with human CCR1 transfectants and not the parental cell line in flow cytometry. No cross-reactivity with CCR5 transfectants is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction