missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR5 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB180S-100UG
This item is not returnable.
View return policy
Description
CCR5 Monoclonal specifically detects CCR5 in Human samples. It is validated for Flow Cytometry.
Specifications
| CCR5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P51681 | |
| CCR5 | |
| NS0 mouse myeloma cell line transfected with human CCR5 Met1-Leu352 Accession # P51681 | |
| RUO | |
| 1234 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 45502 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR5, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Specifically stains human CCR5 transfectants but not the parental cell line as detected by flow cytometry. It has also been shown to react with a CCR5/CCR2 chimera (5222) containing the amino-terminal domain of CCR5. However, this antibody does not react with CCR5 present on unfixed stimulated human PBMCs by flow cytometry. For additional information regarding epitope specificity for this antibody and other R&D Systems anti-human CCR5 antibodies, see Lee,áB. et al., 1999, J.áBiol. Chem. 274:9617. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction