missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR7 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB197RG-100UG
This item is not returnable.
View return policy
Description
CCR7 Monoclonal specifically detects CCR7 in Human samples. It is validated for Flow Cytometry.
Specifications
| CCR7 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| BLR2, BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR7, CCR-7, CD197, CD197 antigen, CDw197, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7, CMKBR7chemokine (C-C) receptor 7, EBI1, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor | |
| Human CCR7 transfectants, Met1-Pro378, Accession # AAA58615 | |
| 100 μg | |
| Primary | |
| Detects human CCR7. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 150503R | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| AAA58615 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 1236 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction