missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD-M6PR Lumenal Domain Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF5320
This item is not returnable.
View return policy
Description
CD-M6PR Polyclonal specifically detects CD-M6PR in Human samples. It is validated for Western Blot.
Specifications
| CD-M6PR | |
| Polyclonal | |
| Biotin | |
| 000 Man6PR, 46 kDa mannose 6-phosphate receptor, 46-kDa mannose 6-phosphate receptor, cation-dependent mannose-6-phosphate receptor, CD Man-6-P receptor, CDM6PR, CD-MPR, FLJ32994, M6PR, mannose-6-phosphate receptor (cation dependent), MPR 46, MPR46, MPRD, Mr 46, SMPR | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4074 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| M6PR | |
| Mouse myeloma cell line NS0-derived recombinant human CD-M6PR Lumenal Domain | |
| 50 μg | |
| Primary | |
| Detects human CD-M6PR Lumenal Domain in Western blots. In Western blots, approximately 40% cross-reactivity with recombinant mouse CD-M6PR is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction