missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD14 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB3832RN-100UG
This item is not returnable.
View return policy
Description
CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD14 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CD14, CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD14, Thr20-Cys352, Accession # P08571 | |
| 100 μg | |
| 929 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 134620R | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| P08571 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human CD14 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction