missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD163 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems FAB11579G-100UG
This item is not returnable.
View return policy
Description
CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for Flow Cytometry
Specifications
| CD163 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Q86VB7 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9332 | |
| Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months, 2 to 8 °C under sterile conditions after opening. | |
| IgG |
| Flow Cytometry | |
| 2977B | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CD_antigen: CD163, CD163, CD163 molecule, GHI/61, HbSR, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, RM3/1, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163 | |
| Mouse myeloma cell line, NS0-derived human CD163, Gly41-Ser1045, Accession # Q86VB7 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction