missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD20 F(ab')2 (Research Grade Ibritumomab Biosimilar) Antibody, Novus Biologicals™

Human Monoclonal Antibody
€ 201.00 - € 600.00
Specifications
| Antigen | CD20 |
|---|---|
| Clone | Hu10F2 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18795765
![]() |
R&D Systems
MAB11054-FAB2-100 |
100 μg |
€ 600.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18736115
![]() |
R&D Systems
MAB11054-FAB2-SP |
25 μg |
€ 201.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow CytometrySpecifications
| CD20 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 931 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Hu10F2 | |
| Flow Cytometry | |
| Unconjugated | |
| Human | |
| Human | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35, Bp35MGC3969, CD20, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Ly-44, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A1, MS4A2, S7 | |
| Human MS4A1/CD20, | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title