missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD24 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5248-SP
This item is not returnable.
View return policy
Description
CD24 Monoclonal specifically detects CD24 in Human samples. It is validated for Immunohistochemistry.
Specifications
| CD24 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P25063 | |
| CD24 | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD24 Ser27-Gly59 (predicted) Accession # P25063 | |
| 25 μg | |
| Primary | |
| Detects human CD24 in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 528807 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD 24, CD24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24A, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 100133941 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction