missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD25/IL-2R alpha Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AB-223-NA
This item is not returnable.
View return policy
Description
CD25/IL-2R alpha Polyclonal specifically detects CD25/IL-2R alpha in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| CD25/IL-2R alpha | |
| Polyclonal | |
| Unconjugated | |
| CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
| Goat | |
| Protein A or G purified | |
| RUO | |
| 3559 | |
| Reconstitute at 1 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 5-20 ug/mL | |
| IL2RA | |
| recombinant human CD25/IL-2 R alpha extracellular domain | |
| 1 mg | |
| Primary | |
| Detects human CD25/IL-2 R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) IL-2 R beta , rhIL-2 R gamma , and rhIL-15 R is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction