missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CD3 epsilon Heterodimer Recombinant Protein
Click to view available options
Quantity:
50 μg
500 μg
Unit Size:
500µg
50µg
Description
It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (200 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.
CD3E and CD3D Heterodimer Protein is produced by mammalian expression system and are involved in mediating signal transduction.
Specifications
Specifications
| Accession Number | P04234, P07766 |
| Concentration | 0.25 mg/mL |
| For Use With (Application) | Control, ELISA Standard, Binding Assay |
| Format | Lyophilized |
| Formulation | PBS with 5% mannitol, 5% trehalose and no preservative; pH 7.4 |
| Gene ID (Entrez) | 915, 916 |
| Molecular Weight (g/mol) | 77.5 kDa |
| Name | Human CD3 epsilon Heterodimer |
| pH Range | 7.4 |
| Purification Method | Purified |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction