Learn More
Human CD35 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB5748-SP
This item is not returnable.
View return policy
Description
CD35 Monoclonal specifically detects CD35 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| CD35 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| CAA68755 | |
| CR1 | |
| Mouse myeloma cell line NS0-derived recombinant human CD35 Gln42-Asp1971 (His1208Arg) Accession # CAA68755 | |
| 25 μg | |
| Primary | |
| Detects human CD35 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant rat CSMD1, recombinant human (rh) CSMD2, rhCSMD3, rhHABP1/C1QBP, or rhComplement Factor H is observed. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 594708 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C3BR, CD35, complement component (3b/4b) receptor 1 (Knops blood group), CR1, KN | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1378 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |