missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD38 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2404V-100UG
This item is not returnable.
View return policy
Description
CD38 Monoclonal specifically detects CD38 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD38 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P28907 | |
| CD38 | |
| NS0 mouse myeloma cell line transfected with human CD38 Met1-Ile300 Accession # P28907 | |
| RUO | |
| 952 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 240742 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| 2'-phospho-ADP-ribosyl cyclase, 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase (EC:2.4.99.20), 2'-phospho-cyclic-ADP-ribose transferase, ADPRC 1, ADPRC1, ADP-ribosyl Cyclase, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1, cADPr hydrolase 1, CD_antigen: CD38, CD38, CD38 antigen, CD38 antigen (p45), CD38 molecule, cluster of differentiation 38, Cyclic ADP-ribose Hydrolase, Cyclic ADP-ribose hydrolase 1, EC 2.4.99.20, EC 3.2.2.6, EC:3.2.2.6, NAD(+) nucleosidase, T10 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CD38. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction