missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD4 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB3791V-025
This item is not returnable.
View return policy
Description
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD4 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 920 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 11830 | |
| Flow Cytometry 5 uL/10^6 cells | |
| P01730 | |
| CD4 | |
| Recombinant human CD4 extracellular domain Accession # P01730 | |
| 25 Tests | |
| Primary | |
| Detects human CD4 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction