missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD4 Antibody, Novus Biologicals™
Human Monoclonal Antibody
€ 248.00 - € 516.00
Specifications
| Antigen | CD4 |
|---|---|
| Clone | 30345-1 |
| Dilution | Cell depletion 2 μg/mL |
| Applications | Depletion Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30241015
|
R&D Systems
MAB11530-100 |
100 μg |
€ 516.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30241043
|
R&D Systems
MAB11530-SP |
25 μg |
€ 248.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Cell depletionSpecifications
| CD4 | |
| Cell depletion 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2μm filtered solution in PBS. | |
| 920 | |
| IgG2 | |
| Protein G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 30345-1 | |
| Depletion Assay | |
| Unconjugated | |
| Human | |
| Human | |
| CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| PHA-stimulated PBMCs | |
| Primary | |
| Reconstitute lyophilized material at 0.2mg/ml in sterile PBS. For liquid material, refer to CoA for concentration. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title