missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD40 Ligand/TNFSF5 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF617-SP
This item is not returnable.
View return policy
Description
CD40 Ligand/TNFSF5 Polyclonal specifically detects CD40 Ligand/TNFSF5 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| CD40 Ligand/TNFSF5 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40L, CD40-L, CD40LG, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5, TNFSF5IMD3, TRAP, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 959 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P29965 | |
| CD40LG | |
| E. coli-derived recombinant human CD40 Ligand/TNFSF5 Glu108-Leu261 Accession # P29965 | |
| 25 μg | |
| Primary | |
| Detects human CD40 Ligand/TNFSF5 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse CD40 Ligand is observed and less than 1% cross-reactivity with recombinant human (rh) CD27 Ligand and rhCD30 Ligand is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction