missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD45 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB14301V-100UG
This item is not returnable.
View return policy
Description
CD45 Monoclonal specifically detects CD45 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD45 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Mouse myeloma cell line NS0-derived recombinant human CD45, Thr24-Asn575, Accession No. P08575 | |
| 100 μg | |
| 5788.0 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 998209 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| P08575 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CD45 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction