Learn More
Human CD6 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB627
 View more versions of this product
Product Code. 18171431
  • € 528.00 / 500µg
Description
CD6 Monoclonal specifically detects CD6 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD6
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD6, CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
923
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Western Blot
123119
Western Blot 1 ug/mL
CAA43306
CD6
Mouse myeloma cell line NS0-derived recombinant human CD6 His18-Glu398 Accession # CAA43306
500 μg
Primary
Detects human CD6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CD6 is observed.