missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB627-SP
This item is not returnable.
View return policy
Description
CD6 Monoclonal specifically detects CD6 in Human samples. It is validated for Western Blot.
Specifications
| CD6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD6, CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 923 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 123119 | |
| Western Blot 1 ug/mL | |
| CAA43306 | |
| CD6 | |
| Mouse myeloma cell line NS0-derived recombinant human CD6 His18-Glu398 Accession # CAA43306 | |
| 25 μg | |
| Primary | |
| Detects human CD6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CD6 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction