missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD7 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB7579G-100UG
This item is not returnable.
View return policy
Description
CD7 Monoclonal specifically detects CD7 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD7 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P09564 | |
| CD7 | |
| Human embryonic kidney cell line HEK293-derived human CD7 Ala26-Pro180 Accession # P09564 | |
| RUO | |
| 924 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 848438 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD7, CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41, TP41p41 protein | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CD7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse CD7 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction