missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD79A Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB69201V-100UG
This item is not returnable.
View return policy
Description
CD79A Monoclonal specifically detects CD79A in Human samples. It is validated for Flow Cytometry.
Specifications
| CD79A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P11912 | |
| CD79A | |
| Mouse myeloma cell line NS0-derived recombinant human CD79A Leu33-Arg143 Accession # P11912 | |
| RUO | |
| 973 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 706931 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD79A, CD79a antigen, CD79A antigen (immunoglobulin-associated alpha), CD79a molecule, immunoglobulin-associated alpha, IGAB-cell antigen receptor complex-associated protein alpha chain, Ig-alpha, MB1, MB-1, MB-1 membrane glycoprotein, Membrane-bound immunoglobulin-associated protein, Surface IgM-associated protein | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CD79A in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse CD79A is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction