missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD79A Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB69201-SP
This item is not returnable.
View return policy
Description
CD79A Monoclonal specifically detects CD79A in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| CD79A | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P11912 | |
| CD79A | |
| Mouse myeloma cell line NS0-derived recombinant human CD79A Leu33-Arg143 Accession # P11912 | |
| 25 μg | |
| Primary | |
| Detects human CD79A in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse CD79A is observed. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 706931 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD79A, CD79a antigen, CD79A antigen (immunoglobulin-associated alpha), CD79a molecule, immunoglobulin-associated alpha, IGAB-cell antigen receptor complex-associated protein alpha chain, Ig-alpha, MB1, MB-1, MB-1 membrane glycoprotein, Membrane-bound immunoglobulin-associated protein, Surface IgM-associated protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 973 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction