missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD79B Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB6620G-100UG
This item is not returnable.
View return policy
Description
CD79B Monoclonal specifically detects CD79B in Human samples. It is validated for Flow Cytometry.
Specifications
| CD79B | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| B29, B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79B, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGB, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD79B, Ala29-Asp159, Accession # P40259 | |
| 100 μg | |
| 974 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 683023 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| P40259 | |
| Mouse | |
| IgM-specific Affinity-purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CD79B in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse CD79B or recombinant human CD79A is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction