Learn More
Human CD8 Antibody, Novus Biologicals™
Human Monoclonal Antibody
Brand:  R&D Systems MAB11531-SP
 View more versions of this product
Product Code. 30241011
  • € 244.00 / 25µg
Description
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Cell depletion
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD8
Monoclonal
Unconjugated
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2μm filtered solution in PBS.
CD_antigen: CD8a, CD8, CD8 antigen, alpha polypeptide (p32), CD8A, CD8a molecule, LEU2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2
Alpha chain of CD8, Accession # P01732
25 μg
Primary
Reconstitute lyophilized material at 0.2mg/ml in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG1
Depletion Assay
30798-1
Cell depletion 2 μg/mL
P01732
Human
Protein G purified from cell culture supernatant
RUO
925