missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD83 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB17741G-100UG
This item is not returnable.
View return policy
Description
CD83 Monoclonal specifically detects CD83 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD83 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| B-cell activation protein, BL11, BL11CD83 antigen, CD83, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15, HB15hCD83 | |
| Human embryonic kidney cell line HEK293-derived recombinant human CD83 Met1-Ala143 Accession # Q01151 | |
| 100 μg | |
| 9308 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 982502 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q01151 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CD83 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction