missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human CD84 / SLAMF5 Protein, His Tag

Product Code. 15846187
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15846187 100 μg 100µg
15856187 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15846187 Supplier ACROBiosystems Supplier No. CD4H5229100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Leukocyte differentiation antigen CD84 is also known as SLAM family member 5 (SLAMF5), which belongs to immunoglobulin (Ig) superfamily. CD84 / SLAMF5 contains one Ig-like C2-type (immunoglobulin-like) domain. CD84 plays a role as adhesion receptor functioning by homophilic interactions and by clustering. CD84 / SLAMF5 increases proliferative responses of activated T-cells and SH2D1A/SAP does not seen be required for this process. Homophilic interactions enhance interferon gamma/IFNG secretion in lymphocytes and induce platelet stimulation via a SH2D1A/SAP-dependent pathway.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.6 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number AAH20063
Molecular Weight (g/mol) 23.6 kDa
Quantity 100 μg
Source Human CD84, His Tag (CD4-H5229) is expressed from human 293 cells (HEK293). It contains AA Lys 22 - Gly 225 (Accession # AAH20063).
Immunogen Lys 22 - Gly 225
Storage Requirements -20°C
Gene Alias CD84,SLAMF5,LY9B,SLAMF5
Common Name CD84
Conjugate Unconjugated
Species Human
Protein Tag His Tag
Buffer PBS, pH7.4
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Immune Checkpoint Proteins
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.