missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD96 v2 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB6199U-100UG
This item is not returnable.
View return policy
Description
CD96 Monoclonal specifically detects CD96 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD96 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_005807 | |
| CD96 | |
| Mouse myeloma cell line NS0-derived recombinant human CD96 Val22-Asp501 Accession # NP_005807 | |
| RUO | |
| 10225 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 628211 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD96, CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILE, TACTILEincreased late expression, T-cell surface protein tactile | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CD96 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction