missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD96v2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6199-SP
This item is not returnable.
View return policy
Description
CD96 Monoclonal specifically detects CD96 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| CD96 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| NP_005807 | |
| CD96 | |
| Mouse myeloma cell line NS0-derived recombinant human CD96 Val22-Asp501 Accession # NP_005807 | |
| 25 μg | |
| Primary | |
| Detects human CD96 in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 628211 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD96, CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILE, TACTILEincreased late expression, T-cell surface protein tactile | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10225 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction