missing translation for 'onlineSavingsMsg'
Learn More

Human CD99 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Product Code. 18759956
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18759956 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18759956 Supplier R&D Systems Supplier No. FAB3968S100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD99 Monoclonal specifically detects CD99 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD99
Applications Flow Cytometry
Classification Monoclonal
Clone 1021527
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P14209
Gene Alias 12E7, CD99, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, pilr-1, PILR-L, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2
Host Species Mouse
Immunogen Mouse myeloma cell line, NS0-derived human CD99, Asp23-Asp122, Accession # P14209
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4267
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.