missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CD99L2 (aa 27-117) Control Fragment Recombinant Protein

Product Code. 30208262
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30208262

Brand: Invitrogen™ RP95695

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (51%), Rat (51%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58539 (PA5-58539. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CD99L2 may function as a homophilic adhesion molecule. CD99L2 functions in leukocyte-endothelial cell interactions during leukocyte extravasation, and in particular, at the diapedesis step. CD99L2 does not seem to be involved in docking of leukocytes to the vessel wall or in lymphocyte diapedesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8TCZ2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 83692
Name Human CD99L2 (aa 27-117) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AW548191; CD99; Cd99 antigen-like 2; CD99 antigen-like protein 2; CD99 molecule like 2; CD99 molecule-like 2; CD99B; CD99L2; MIC2 (monoclonal Imperial Cancer Research Fund 2)-like 1; MIC2 like 1; MIC2 like-1; MIC2L1; MIC2-like protein 1; protein rhombex-40; Rhombencephalic expression protein 40 kDa; Rhombe x 40; Rhombex-40; single-pass transmembrane protein; UNQ1964/PRO4486; vms-tm2; Xap89
Common Name CD99L2
Gene Symbol Cd99l2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FDDFNLEDAVKETSSVKQPWDHTTTTTTNRPGTTRAPAKPPGSGLDLADALDDQDDGRRKPGIGGRERWNHVTTTTKRPVTTRAPANTLGN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.