missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CEACAM-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4354-SP
This item is not returnable.
View return policy
Description
CEACAM4 Monoclonal specifically detects CEACAM4 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CEACAM4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| O75871 | |
| CEACAM4 | |
| E. coli-derived recombinant human CEACAM-4 Phe36-Val140 Accession # O75871 | |
| 25 μg | |
| Primary | |
| Detects human CEACAM-4 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CEACAM-1, -3, -5, -6, -7, or -8 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 568410 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| carcinoembryonic antigen CGM7, carcinoembryonic antigen gene family member 7, carcinoembryonic antigen-related cell adhesion molecule 4, CEACAM4, CGM7, CGM7_HUMAN | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1089 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction