missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CEACAM-7 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB44781T-100UG
This item is not returnable.
View return policy
Description
CEACAM7 Monoclonal specifically detects CEACAM7 in Human samples. It is validated for Flow Cytometry.
Specifications
| CEACAM7 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Carcinoembryonic antigen CGM 2, Carcinoembryonic antigen CGM2, Carcinoembryonic antigen gene family member 2, Carcinoembryonic antigen related cell adhesion molecule 7, CEA, CEACAM 7, CEACAM7, CGM 2, CGM2 | |
| Chinese Hamster Ovary cell line, CHO-derived recombinant human CEACAM-7 Thr36-Asn233 Accession # Q14002 | |
| 100 μg | |
| 1087 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 962703 | |
| Flow Cytometry 0.25-1μg/106 cells | |
| Q14002 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CEACAM-7 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction