missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CEACAM-7 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems BAF4478
This item is not returnable.
View return policy
Description
CEACAM7 Polyclonal specifically detects CEACAM7 in Human samples. It is validated for Western Blot.
Specifications
| CEACAM7 | |
| Polyclonal | |
| Biotin | |
| Q14002 | |
| CEACAM7 | |
| E. coli-derived recombinant human CEACAM-7 Asn37-Phe142 Accession # Q14002 | |
| 50 μg | |
| Primary | |
| Detects human CEACAM-7 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant human CEACAM-1, -3, -4, -5, -6, and -8 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Carcinoembryonic antigen CGM 2, Carcinoembryonic antigen CGM2, Carcinoembryonic antigen gene family member 2, Carcinoembryonic antigen related cell adhesion molecule 7, CEA, CEACAM 7, CEACAM7, CGM 2, CGM2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1087 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction