missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CEACAM-8/CD66b Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems BAF4246
This item is not returnable.
View return policy
Description
CEACAM8/CD66b Polyclonal specifically detects CEACAM8/CD66b in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| CEACAM8/CD66b | |
| Polyclonal | |
| Biotin | |
| P31997 | |
| CEACAM8 | |
| E. coli-derived recombinant human CEACAM-8/CD66b Gln35-His141 Accession # P31997 | |
| 50 μg | |
| Primary | |
| Detects human CEACAM-8/CD66b in Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) CEACAM-3 and rhCEACAM-7 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
| Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CEACAM8, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1088 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction