missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CHML (aa 104-168) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (48%), Rat (48%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56213 (PA5-56213. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Binds unprenylated Rab proteins, presents it to the catalytic component B, and remains bound to it after the geranylgeranyl transfer reaction. The component A may be regenerated by transferring its prenylated Rab to a protein acceptor. Less effective than REP-1 in supporting prenylation of Rab3 family.
Specifications
Specifications
| Accession Number | P26374 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 1122 |
| Name | Human CHML (aa 104-168) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AI450814; CHM like, Rab escort protein 2; Chml; choroideraemia-like protein; choroideremia-like; choroideremia-like (Rab escort protein 2); choroideremia-like protein; E030003F13Rik; rab escort protein 2; rab escort protein-2; rab proteins geranylgeranyltransferase component A 2; REP2; REP-2 |
| Common Name | CHML |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction