missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CHN2 (aa 130-221) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-139765 (PA5-139765. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene is a member of the chimerin family and encodes a protein with a phorbol-ester/DAG-type zinc finger, a Rho-GAP domain and an SH2 domain. This protein has GTPase-activating protein activity that is regulated by phospholipid binding and binding of diacylglycerol (DAG) induces translocation of the protein from the cytosol to the Golgi apparatus membrane. The protein plays a role in the proliferation and migration of smooth muscle cells. Decreased expression of this gene is associated with high-grade gliomas and breast tumors, and increased expression of this gene is associated with lymphomas. Mutations in this gene have been associated with schizophrenia in men. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
| Accession Number | P52757 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 1124 |
| Name | Human CHN2 (aa 130-221) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1700026N20Rik; 4930557O16Rik; ARHGAP3; BCH; beta chimaerin; beta chimerin; beta3-chimaerin; beta-chimaerin; Beta-chimerin; chimerin (chimaerin) 2; chimerin 2; chimerin, beta-2; chimerin, testis-specific; Chn2; CHN2-3; OTTHUMP00000202260; Rho GTPase-activating protein 3; RHOGAP3; rho-GTPase-activating protein 3 |
| Common Name | CHN2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction