missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chondrolectin Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2576V-100UG
This item is not returnable.
View return policy
Description
Chondrolectin Monoclonal specifically detects Chondrolectin in Human samples. It is validated for Flow Cytometry.
Specifications
| Chondrolectin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9H9P2 | |
| CHODL | |
| Mouse myeloma cell line NS0-derived recombinant human Chondrolectin Arg22-Asn216 Accession # Q9H9P2 | |
| RUO | |
| 140578 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 396305 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| C21orf68, CHODL, chondrolectin, chromosome 21 open reading frame 68, FLJ12627, MT75, PRED12, Transmembrane protein MT75 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Chondrolectin in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction