missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CHRM3 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB6378U-100UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Muscarinic Acetylcholine Receptor M3/CHRM3 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M3/CHRM3 in Human samples. It is validated for Flow Cytometry
Spécification
| Muscarinic Acetylcholine Receptor M3/CHRM3 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| cholinergic receptor, muscarinic 3, HM3, m3 muscarinic receptor, mAChR M3, muscarinic acetylcholine receptor M3 | |
| NS0 mouse myeloma cell line transfected with human CHRM3, Accession # P20309 | |
| 100 μg | |
| Primary | |
| Reconstitution default not known | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 580011 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P20309 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1131 | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu