missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CILP-1 C-Terminal Fragment Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4477-SP
This item is not returnable.
View return policy
Description
CILP-1 Polyclonal specifically detects CILP-1 in Human samples. It is validated for Western Blot.
Specifications
| CILP-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cartilage intermediate layer protein 1, cartilage intermediate layer protein 1 C1, cartilage intermediate layer protein 1 C2, cartilage intermediate layer protein, nucleotide pyrophosphohydrolase, Cartilage intermediate-layer protein, CILP, CILP1, CILP-1, HsT18872 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 8483 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| O75339 | |
| CILP | |
| Mouse myeloma cell line NS0-derived recombinant human CILP-1 C-Terminal Fragment Arg724-Asn1184 Accession # O75339 | |
| 25 μg | |
| Primary | |
| Detects human CILP-1 C-Terminal Fragment in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human CILP-1 N-terminal peptide is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction